ICI Quick Ship Patriot Table Complete Plumbing Assembly, Vacuum; Yellow
Representative image only.
ICI Quick Ship Patriot Table Complete Plumbing Assembly, Vacuum; Yellow
ICI Quick Ship | Mfr: CPPTCPA-V| Item: EW-78983-07
Enhance the capabilities of your ICI Quick Ship Patriot Tables with these accessories
$538.00USD / Each
$538.00USD / Each
Product Description
Includes
ball valve service turret with shank, a service hose, and a male quick connect fitting.
Key Features
  • Chose accessories that are compatible with your Patriot table type and width
  • Features a single end shut off plug and does not have an internal valve
  • For use with vacuum service
  • 2 year warranty
More About this Item
Optional accessories include shelving, lighting, electrical outlets, plumbing assemblies, and more. Choose from a wide range of Patiot table options, which are sold separately.
All quick connects have 316 stainless steel internal components. Keyed quick connect members will only couple with components of the same key which are identified by a color code. Assemblies consist of 2 major components and are shipped loose to be installed on site. The service hoses carry services from the overhead service panel, through the upright of the table and to the shank of the fixture. They consist of color coded reinforced PVC 1/4” hose. The ball valve is a polished chrome valve with a brass floating cone and integral brass seat. They provide metering control of laboratory gases at pressures up to 60 PSI. The turret shank is bored for the connection of the service hose.
Two year warranty
Specifications
  • Material316 stainless steel quick connects, PVC hose, and polished chrome ball valve with a brass floating cone and integral brass seat
  • DescriptionPatriot Table Complete Plumbing Assembly, Vacuum; Yellow
  • Warranty2 Years