Product Type
In Stock
Max Pressure (in WC)
Min Pressure (in WC)
Industry
Max Pressure (PSID)
Regulatory Compliance
US Origin
Pressure Transducers
The product searched is discontinued, please browse below for replacements.
Pressure Transducers
Pressure transducers convert an applied pressure through a process connection into a current or voltage output. Traditionally, pressure transmitters have a current (4 to 20 mA) output, while pressure transducers have a voltage output, but the terminology is interchangeable when talking about pressure instruments. All pressure transmitters and pressure transducers are used for their output to control pumps, switches, PLCs, or just send a signal to a panel meter to control processes or signal a need for attention. We have sanitary, industrial, differential, and barometric pressure transducers to meet your needs.
APPLY FILTERS
filter byclear all
42 RESULTS
SORT BY
Showing 1 - 24 of 42
2>