Justrite 8508 Steel Drum Lock Set
Representative image only.
Justrite 8508 Steel Drum Lock Set
Justrite | Mfr: 08508| Item: EW-81792-61
Safely secure drum contents without the risk of contamination
$136.50USD / Set of 1
$136.50USD / Set of 1
Product Description
Includes
One 3/4" NPT and one 2" NPT lock (padlocks not included).
Key Features
  • Install in just seconds
  • Fits most popular styles of steel and plastic 55-gallon drums
  • Each set contains two locks
More About this Item
Justrite drum locks are a cost-effective solution to controlling storage concerns of all types of liquids, semi-liquids, and powders that come in standard drums. These drum locks fit the most popular style of steel and plastic 55-gallon (200L) drums. The locks are corrosion resistant, non-sparking zinc die-cast. They install over bung flange — no potential for content contamination.
Specifications
  • DescriptionSteel Drum Lock Set