Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 10, 60/bag
Representative image only.
Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 10, 60/bag
Kimberly-Clark | Mfr: 11828| Item: EW-86485-36
Contain no latex proteins for sensitive workers
$90.90USD / Bag of 60
BUY MORE & SAVE
  • 1-9$90.90 USD / Bag of 60
  • 10+$81.81 USD / Bag of 60
$90.90USD / Bag of 60
BUY MORE & SAVE
  • 1-9$90.90 USD / Bag of 60
  • 10+$81.81 USD / Bag of 60
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Kimtech Pure® cleanroom gloves are recommended for use in ISO Class 3 cleanroom or higher. These disposable gloves are made of a synthetic nitrile polymer and contain no natural rubber latex or silicone. Gloves feature textured fingertips for added grip, beaded cuffs for ease of donning, and a smooth finish to accommodate double donning. Each package includes sixty gloves, double-bagged in sealed cleanroom bags. Ambidextrous.
Compliance: ISO Class 3
Specifications
  • Thickness (mil)4
  • MaterialNitrile
  • Size10
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • ComplianceISO Class 3
  • SterileNo
  • DescriptionNitrile Cleanroom Gloves, Size 10, 60/bag