Lab-Crest 110-858-0001 Coupling Component, neoprene o-ring
Representative image only.
Lab-Crest 110-858-0001 Coupling Component, neoprene o-ring
Lab-Crest | Mfr: 110-858-0001| Item: EW-63400-70
$4.90USD / Each
$4.90USD / Each
Buy It Together
Current Item
Lab-Crest 110-858-0001 Coupling Component, neoprene o-ring
Lab-Crest 110-858-0001 Coupling Component, neoprene o-ring
6340070
$4.90USD / Each
+ =
Total Price 257.65 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Parts and accessories for your Lab-Crest system.
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an o-ring.
This is the Neoprene® version of the needed o-ring.
Specifications
  • MaterialNeoprene
  • Compatible WithLab-Crest Pressure Reaction Vessels
  • DescriptionCoupling Component, neoprene o-ring