LabChem Murexide Indicator for Calcium, Dry Form in NaCl; 500 g

Representative image only.

LabChem Murexide Indicator for Calcium, Dry Form in NaCl; 500 g

LabChem | Mfr: LC175101| Item: EW-80029-39

Highest quality raw materials!

$310.50USD / Each

$310.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 3500-Ca B
  • Purity Grade
  • MediumSodium Chloride
  • Element/IonCalcium
  • Weight (g)500
  • PackagingPoly Bottle
  • Shelf Life (days)1095
  • Shelf Life (months)36
  • DescriptionMurexide Indicator, Dry Form in NaCl, for Calcium; 500 g
Reviews
Q&A