LabChem Nitrate (as Nitrogen) Standard, 1000 ppm; 125 mL

Representative image only.

LabChem Nitrate (as Nitrogen) Standard, 1000 ppm; 125 mL

LabChem | Mfr: LC179007| Item: UX-80029-84

Highest quality raw materials!

$39.80USD / Each

$39.80USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 4500-NO₃ˉ E, ASTM D3867-04 A,B
  • Concentration1000 ppm N, 4430 ppm NO₃ˉ
  • Purity Grade
  • ChemistryStandards
  • Element/IonNitrate
  • Volume (mL)125
  • PackagingPoly Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionNitrate (as Nitrogen) Standard, 1000 ppm, (1 mL=1 mg N, 1 mL=4.43 mg NO₃ˉ); 125 mL
Reviews
Q&A