LabChem Nitrate (as Nitrogen) Standard
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$39.80 - $68.15USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Nitrate (as Nitrogen) Standard, 1000 ppm; 500 mL EW-80029-81 Mfr # LC179001
COMPARE
500 Nitrate (as Nitrogen) Standard, 1000 ppm, (1 mL=1 mg N, 1 mL=4.43 mg NO₃ˉ); 500 mL
$68.15USD / Each
LabChem Nitrate (as Nitrogen) Standard, 1000 ppm; 125 mL EW-80029-84 Mfr # LC179007
COMPARE
125 Nitrate (as Nitrogen) Standard, 1000 ppm, (1 mL=1 mg N, 1 mL=4.43 mg NO₃ˉ); 125 mL
$39.80USD / Each