LabChem Phosphate Buffer pH 7.2 for BOD; 4 L

Representative image only.

LabChem Phosphate Buffer pH 7.2 for BOD; 4 L

LabChem | Mfr: LC185004| Item: EW-80031-71

Highest quality raw materials!

$137.00USD / Each

$137.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 5210 B
  • Purity Grade
  • ChemistrySolutions
  • Element/IonBOD
  • Volume (mL)4000
  • PackagingPoly Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionPhosphate Buffer pH 7.2, for BOD; 4 L
Reviews
Q&A