LabChem Silver Nitrate, 0.1 N (0.1 M); 4 L

Representative image only.

LabChem Silver Nitrate, 0.1 N (0.1 M); 4 L

LabChem | Mfr: LC227004| Item: EW-80037-05

Highest quality raw materials!

$542.00USD / Each

$542.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.1
  • Purity Grade
  • Concentration0.1 M
  • ChemistrySolutions
  • Volume (mL)4000
  • PackagingAmber Glass Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionSilver Nitrate, 0.1 N (0.1 M); 4 L
Reviews
Q&A