LabChem Sodium Hydroxide, 1.0 N (1.0 M); 1 L

Representative image only.

LabChem Sodium Hydroxide, 1.0 N (1.0 M); 1 L

LabChem | Mfr: LC243502| Item: UX-80044-64

Highest quality raw materials!

$44.90USD / Each

$44.90USD / Each

Buy It Together

Current Item

LabChem Sodium Hydroxide, 1.0 N (1.0 M); 1 L
LabChem Sodium Hydroxide, 1.0 N (1.0 M); 1 L
8004464

$44.90USD / Each
+ + =
Total Price 188.60 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality1
  • Concentration1.0 M
  • Purity Grade
  • ChemistrySolutions
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationClass 8, Packing Group II
  • UN NumberUN1824
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionSodium Hydroxide, 1.0 N (1.0 M); 1 L
Reviews
Q&A