Spectrum Chemical Phenolphthalein 0.5% (w/v) Indicator Solution in 50% Alcohol, APHA; 1 L

Representative image only.

Spectrum Chemical Phenolphthalein 0.5% (w/v) Indicator Solution in 50% Alcohol, APHA; 1 L

Spectrum Chemical | Mfr: P-150-1LT| Item: EW-80161-72

Quality and delivery you can count on every time!

$99.90USD / Each

$99.90USD / Each

Buy It Together

Current Item

Spectrum Chemical Phenolphthalein 0.5% (w/v) Indicator Solution in 50% Alcohol, APHA; 1 L
Spectrum Chemical Phenolphthalein 0.5% (w/v) Indicator Solution in 50% Alcohol, APHA; 1 L
8016172

$99.90USD / Each
+
LabChem Sodium Hydroxide, 1.0 N (1.0 M); 500 mL
8004383

$34.30USD / Each
+
LabChem Hydrochloric Acid, 1.0 N (1.0 M); 1 L
8001958

$44.60USD / Each
+
LabChem Hydrochloric Acid, 0.1 N (0.1 M); 500 mL
8001891

$43.50USD / Each
=
Total Price 222.30 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • ClassUN1170, Ethyl alcohol solution, 3, PG III
  • Volume (mL)1000
  • PackagingPoly Bottle
  • UN NumberUN1170
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionPhenolphthalein 0.5% (w/v) Indicator Solution in 50% Alcohol, APHA; 1 L
Documents & Certificates
Reviews
Q&A