LabChem TISAB II, for Fluoride; 4 L

Representative image only.

LabChem TISAB II, for Fluoride; 4 L

LabChem | Mfr: LC261304| Item: EW-80054-49

Highest quality raw materials!

$274.45USD / Each

$274.45USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 4500-Fˉ C
  • Purity Grade
  • ChemistrySolutions
  • Element/IonFluoride
  • Volume (mL)4000
  • PackagingPoly Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionTISAB II, for Fluoride; 4 L
Documents & Certificates
Reviews
Q&A