Labconco 9019200 WaterPro BT Filter Pack; 1/Pk
Representative image only.
Labconco 9019200 WaterPro BT Filter Pack; 1/Pk
Labconco | Mfr: 9019200| Item: EW-99290-48
Optimize performance of your Labconco WaterPro BT Water Purification Systems
$1,040.00USD / Each
$1,040.00USD / Each
Product Description
Key Features
  • Compatible with the Labconco WaterPro BT Water Purification Systems
  • Required for use of, and with, any Labconco WaterPro BT System
  • 1 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your Labconco WaterPro BT Water Purification Systems.
All water filtration technologies (activated carbon filtration, deionization, and RO) are combined into this one convenient twin-cartridge pack—simply snaps into place!
1 Year Warranty
Specifications
  • DescriptionWaterPro BT Filter Pack; 1/Pk