Labconco FreeZone Fast-Freeze Flask, Amber; 300 mL
Representative image only.
Labconco FreeZone Fast-Freeze Flask, Amber; 300 mL
Labconco | Mfr: 7542601| Item: EW-03353-28
$193.00USD / Each
$193.00USD / Each
Buy It Together
Current Item
Labconco FreeZone Fast-Freeze Flask, Amber; 300 mL
Labconco FreeZone Fast-Freeze Flask, Amber; 300 mL
0335328
$193.00USD / Each
+ +
Labconco FreeZone Standard Freeze Drying Chamber, 12 Ports, SS
0334601
$2,709.00USD / Each
$3,010.00USD / Each
=
Total Price 3312.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Withstands extreme temperatures and high vacuum
  • Amber-colored glass protects light-sensitive samples
  • Flat bottom and wide mouth design provides easy loading and cleaning
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
Specifications
  • Capacity (Liters)0.3
  • Height (InchShort)5 53/64
  • DescriptionFast-Freeze Flask, Amber, Flat Bottom, 300 mL