Labconco FreeZone Five-shelf Sample Stand; 17-1/2” x 10 3/8”
Representative image only.
Labconco FreeZone Five-shelf Sample Stand; 17-1/2” x 10 3/8”
Labconco | Mfr: 7317200| Item: EW-03353-20
$1,453.50USD / Each
$1,615.00USD / Each
$1,453.50USD / Each
$1,615.00USD / Each
Buy It Together
Current Item
Labconco FreeZone Five-shelf Sample Stand; 17-1/2” x 10 3/8”
Labconco FreeZone Five-shelf Sample Stand; 17-1/2” x 10 3/8”
0335320
$1,453.50USD / Each
$1,615.00USD / Each
+ +
Labconco FreeZone Standard Freeze Drying Chamber, 12 Ports, SS
0334601
$2,709.00USD / Each
$3,010.00USD / Each
=
Total Price 4572.50 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
three rubber feet.
Key Features
  • Safely accommodates bulk samples, microtiter plates, vials, and serum bottles
  • Removable shelf design offers flexible height spacing and quick loading
  • Compatible with tall clear freeze drying chambers with or without valves
  • 1 year warranty
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
1 Year Warranty
Specifications
  • Width (InchShort)10 53/64
  • Height (InchShort)17 1/2
  • DescriptionFive-shelf Sample Stand, 17 1/2” x 10 3/8”