Labconco FreeZone Glass Adapter, Straight; 1/2" to 3/4"
Representative image only.
Labconco FreeZone Glass Adapter, Straight; 1/2" to 3/4"
Labconco | Mfr: 7545400| Item: EW-03354-02
$36.10USD / Each
$36.10USD / Each
Buy It Together
Current Item
Labconco FreeZone Glass Adapter, Straight; 1/2" to 3/4"
Labconco FreeZone Glass Adapter, Straight; 1/2" to 3/4"
0335402
$36.10USD / Each
+ +
Labconco FreeZone Standard Freeze Drying Chamber, 12 Ports, SS
0334601
$2,709.00USD / Each
$3,010.00USD / Each
=
Total Price 3155.10 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with 40, 80, 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze Flask Tops
  • 1 year warranty
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
1 Year Warranty
Specifications
  • DescriptionAccessory Straight glass adapter 1/2" to 3/4"