Labconco FreeZone Tube Rack, Round; 104 x 12-13 mm
Representative image only.
Labconco FreeZone Tube Rack, Round; 104 x 12-13 mm
Labconco | Mfr: 7379000| Item: EW-03353-23
$306.00USD / Each
$340.00USD / Each
$306.00USD / Each
$340.00USD / Each
Buy It Together
Current Item
Labconco FreeZone Tube Rack, Round; 104 x 12-13 mm
Labconco FreeZone Tube Rack, Round; 104 x 12-13 mm
0335323
$306.00USD / Each
$340.00USD / Each
+ +
Labconco FreeZone Standard Freeze Drying Chamber, 12 Ports, SS
0334601
$2,709.00USD / Each
$3,010.00USD / Each
=
Total Price 3425.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
stainless steel bottom.
Key Features
  • Securely holds up to one-hundred-four 12 to 13 mm dia test tubes in upright position
  • Compatible with FreeZone heated or non-heated round Product Shelves, Bulk Tray Dryers, Stoppering Tray Dryers, and Tray Dryers
  • 1 year warranty
More About this Item
Adapt, expand capacity, or just keep your freeze dryer working at its best with these parts and accessories.
1 Year Warranty
Specifications
  • Height (InchShort)2 1/2
  • DescriptionTest Tube Rack, Round, Stainless Steel, 12 to 13 mm