LaMotte 4497 Liquid Sanitizer Strength Verification Kits, Chlorine Test
Representative image only.
LaMotte 4497 Liquid Sanitizer Strength Verification Kits, Chlorine Test
LaMotte | Mfr: 4497-01| Item: EW-99532-04
Titrimetric kits verify the concentration of your sanitizer quickly and easily
$77.80USD / Each
$86.45USD / Each
$77.80USD / Each
$86.45USD / Each
Product Description
Key Features
  • Save money by avoiding overuse or underuse of sanitizers
  • 1 year warranty
More About this Item
Verify the strength and effectiveness of your liquid sanitizer with these colorimetric test kits. All kits contain reagents for 50 tests.
1 Year Warranty
Specifications
  • TestChlorine
  • Measurement Points10 ppm/drop
  • Tests Per Kit50
  • DescriptionLiquid Sanitizer Strength Verification Kits, Chlorine Test