Lascar Electronics CABLE USB A-MF USB Interface Cable for Power and Data Transfer
Representative image only.
Lascar Electronics CABLE USB A-MF USB Interface Cable for Power and Data Transfer
Lascar Electronics | Mfr: CABLE USB A-MF| Item: EW-18000-58
Optimize performance of your Lascar Process Control Units
$14.10USD / Each
$14.10USD / Each
Buy It Together
Current Item
Lascar Electronics CABLE USB A-MF USB Interface Cable for Power and Data Transfer
Lascar Electronics CABLE USB A-MF USB Interface Cable for Power and Data Transfer
1800058
$14.10USD / Each
+ =
Total Price 40.20 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Keep your running properly with these cables and adapters
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Lascar Process Control meters
USB cable; Type A to mini B for connection of a PanelPilot compatible display to a Windows PC.
See 18000-59 for connection to a standard North American wall socket for powering a PanelPilot display.
Does not provide a waterproof connection to the 18000-23 IP67 rated PanelPilot graphic panel meter. However, it is needed for programming and data transfer.
1 Year Warranty
Specifications
  • Compatible With18000-17, 18000-18, 18000-19, 18000-20, 18000-21, 18000-22, 18000-23
  • Length (cm)200
  • DescriptionUSB Interface Cable for Power and Data Transfer