Lovibond RS5550 Colorimeter Reagent Set, Total Nitrogen, Low Range, 50 tests
Representative image only.
Lovibond RS5550 Colorimeter Reagent Set, Total Nitrogen, Low Range, 50 tests
Lovibond | Mfr: 535550| Item: EW-99560-75
Premeasured reagents in convenient powder packs
$242.55USD / Pkg of 1
$242.55USD / Pkg of 1
Product Description
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.
Specifications
  • Test ParametersTotal nitrogen, low range
  • Number of Tests Included50
  • DescriptionColorimeter Reagent Set, Total Nitrogen, Low Range, 50 tests