M-System B5MS Two-Wire Transmitter, 100-10k ohm potentiometer in, 4-20 mA out
Representative image only.
M-System B5MS Two-Wire Transmitter, 100-10k ohm potentiometer in, 4-20 mA out
M-System | Mfr: B5MS| Item: EW-68467-63
$149.17USD / Each
$175.50USD / Each
$149.17USD / Each
$175.50USD / Each
Product Description
Key Features
  • 3 year warranty
More About this Item
M-System's M5 / B5 / W5 Series signal conditioners are designed to accept a wide variety of process signal input and provide a standard and non-standard DC output. Only 1.6'' (41mm) deep modules can be installed anywhere, even behind the panel cover.
Th M5 series features signal channel four-wire models, while the B5 series are two-wire models. Model B5SN input loop powered isolator can tap into the existing 4-20mA loops without needing to add additional wiring for power. Model B5VS 2-wire output loop powered isolator is designed to accept a DC voltage or current signal input and provide a 4-20mA output. The W5 series signal splittes provide two isolated DC outputs of independent ranges.
3 Year Warranty
Specifications
  • Input type100Ω to 10kΩ Potentiometer
  • DescriptionTwo-Wire Transmitter, 100-10k ohm potentiometer in, 4-20 mA out
  • Warranty3 Years