M-System M2DYS-24A-M2 Isolated Curnt Loop Supply 24V/22mA 4-20 Ma 4-20 Ma100 - 240V Ac

Representative image only.

M-System M2DYS-24A-M2 Isolated Curnt Loop Supply 24V/22mA 4-20 Ma 4-20 Ma100 - 240V Ac

M-System | Mfr: M2DYS-24A-M2/UL| Item: EW-68467-44

$215.47USD / Each
$253.50USD / Each

$215.47USD / Each
$253.50USD / Each
Product Description

Key Features

  • Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring
  • 2000 VAC Isolation
  • Standard DIN rail or surface mounting
  • Base socket included with modules
  • CE marking and UL/C-UL Class I Div 2 approval
  • 3 year warranty

More About this Item

M-System's Mini-M Series Signal Conditioners are designed to accept the largest range of process signal inputs and provide a standard and non-standard DC output.

The Mini-MW Series Signal Splitters provide a second isolated output of independent range, which give you the flexibility to add sophisticated distributed control or MIS monitoring to a local loop, without a worry about mismatched impedance, or the threat of one problem system's output signal impacting the other.

Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring.

3 Year Warranty
Specifications
  • Max Input (mA)20
  • Min Input (mA)4
  • Max Output ch1 (mA)20
  • Min Output ch1 (mA)4
  • Power Supply (VAC)100 to 240
  • Max Operating Temperature (° C)55
  • Min Operating Temperature (° C)-5
  • Max Operating Temperature (° F)131
  • Min Operating Temperature (° F)23
  • Height (in)2 63/64
  • Width (in)29/32
  • Depth (in)4 7/8
  • Height (cm)7.6
  • Width (cm)2.3
  • Depth (cm)12.4
  • DescriptionIsolated Curnt Loop Supply 24V/22mA 4-20 Ma 4-20 Ma100 - 240V Ac
  • Warranty3 Years
Reviews
Q&A