Mark-10 Speed Extension Package for Test Frames with EasyMESUR® Control Panel
Representative image only.
Mark-10 Speed Extension Package for Test Frames with EasyMESUR® Control Panel
Mark-10 | Mfr: EMP002-1| Item: EW-67801-15
Enhance your digital force tester with EasyMESUR® control panel with advanced applications
$625.00USD / Each
sign in for your price.
$625.00USD / Each
sign in for your price.
Product Description
Key Features
  • Field upgradeable—no need to ship it to the factory or call for service
  • Upgrade test frames 67801-10 through -13
  • Includes upgrades for high-speed extension and low-speed extension
  • 3 year warranty
More About this Item
Bundled software packages offer convenience. Optional functions include, distance measurement and limits, speed extension, break detection, load holding, and cycling. Software can be added at any time and easily upgraded in the field.
High-speed extension: Extend the high end of the speed range to 1,100 mm/min (45 inches/min).
Low-speed extension: Extend the low end of the speed range to 0.5 mm/min (0.02 inches/min)
Specifications
  • Compatible WithTest frames with EasyMESUR® control panel 67801-10 through -13
  • DescriptionSpeed Extension Package for Test Frames with EasyMESUR® Control Panel
  • Warranty3 years