Megger 242021-7 Potential Test Lead, Single Hand Spike, 7 ft

Representative image only.

Megger 242021-7 Potential Test Lead, Single Hand Spike, 7 ft

Megger | Mfr: 242021-7| Item: EW-20011-03

$791.60USD / Each
sign in for your price.

$791.60USD / Each
sign in for your price.
Product Description

Key Features

  • 1 year warranty

More About this Item

Select the test lead set(s) for your application.

1 Year Warranty
Specifications
  • Length (in)84
  • Length (cm)200
  • DescriptionPotential Test Lead, Single Hand Spike, 7 ft
  • Warranty1 year
Reviews
Q&A