Mettler Toledo 11106883 ErgoClip Titration Basket Titration Container, Max 50mm for Excellence Analytical Balance
Representative image only.
Mettler Toledo 11106883 ErgoClip Titration Basket Titration Container, Max 50mm for Excellence Analytical Balance
Mettler Toledo | Mfr: 11106883| Item: EW-11336-41
$162.50USD / Each
$162.50USD / Each
Buy It Together
Current Item
Mettler Toledo 11106883 ErgoClip Titration Basket Titration Container, Max 50mm for Excellence Analytical Balance
Mettler Toledo 11106883 ErgoClip Titration Basket Titration Container, Max 50mm for Excellence Analytical Balance
1133641
$162.50USD / Each
+ =
Total Price 325.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Holds titration beaker or test tubes up to 50 mm dia
  • Allows for one step dosing directly into tare container
  • 3 month warranty
More About this Item
Parts and accessories to expand your capabilities.
3 Month Warranty
Specifications
  • DescriptionErgoClip Titration Basket Tare Container, Max 50 mm for Excellence Analytical Balance