Mettler Toledo 30705122 Cuvette Carousel XPathHolder™, 10/16mm
Representative image only.
Mettler Toledo 30705122 Cuvette Carousel XPathHolder™, 10/16mm
Mettler Toledo | Mfr: 30705122| Item: EW-83700-83
Optimize performance of your Mettler Toledo EasyPlus spectrophotometers
$47.50USD / Each
$47.50USD / Each
Product Description
Key Features
  • Expand analysis capabilities and make testing easier
  • Cuvette carousel with cuvette holder is suitable for use with 10 mm cuvette and 16 mm OD glass tube
  • Only one carousel can be installed in the analysis compartment at a time
More About this Item
Choose from a wide range of accessories including cuvette carousels, replacement bulbs, and more for your Mettler Toledo EasyPlus spectrophotometers.
Cuvette holders provide a secure base for your sample, ensuring highly accurate measurement results.
Specifications
  • Pathlength (mm)10 and 16 mm
  • Materials of ConstructionPA66 Polyamide
  • DescriptionCuvette Carousel XPathHolder™, 10/16mm