Mettler Toledo 30705148 Easy VIS Tungsten Lamp Set
Representative image only.
Mettler Toledo 30705148 Easy VIS Tungsten Lamp Set
Mettler Toledo | Mfr: 30705148| Item: EW-83700-81
Optimize performance of your Mettler Toledo EasyPlus spectrophotometers
$156.50USD / Each
$156.50USD / Each
Product Description
Key Features
  • Expand analysis capabilities and make testing easier
  • Easy-to-replace lamp module for self-maintenance
More About this Item
Choose from a wide range of accessories including cuvette carousels, replacement bulbs, and more for your Mettler Toledo EasyPlus spectrophotometers.
The Easy VIS spectrophotometer constantly monitors the lamp and alerts the user when it needs replaced, which only takes two minutes.
Specifications
  • Measurement ModeVisible Spectrophotometer
  • Source LampTungsten lamp
  • Source Lamp Life2000 hours
  • DescriptionEasy VIS Tungsten Lamp Set