Mettler Toledo 30705183 SQKitReader™ Internal Barcode Reader for Spectroquant® Water Testing
Representative image only.
Mettler Toledo 30705183 SQKitReader™ Internal Barcode Reader for Spectroquant® Water Testing
Mettler Toledo | Mfr: 30705183| Item: EW-83700-80
Optimize performance of your Mettler Toledo EasyPlus spectrophotometers
$585.00USD / Each
$585.00USD / Each
Product Description
Key Features
  • Expand analysis capabilities and make testing easier
  • Internal barcode reader reads 2D barcodes of water test kits
More About this Item
Choose from a wide range of accessories including cuvette carousels, replacement bulbs, and more for your Mettler Toledo EasyPlus spectrophotometers.
SQKitReader™ barcode reader scans Spectroquant™ water test kit barcodes to automatically start methods inputted on the user-friendly touchscreen.
Specifications
  • Measurement ModeSpectroquant™ water testing methods
  • OutputPogo Pin
  • DescriptionSQKitReader™ Internal Barcode Reader for Spectroquant® Water Testing