Mettler Toledo EasyPlus Titrator Burette, 50 mL

Representative image only.

Mettler Toledo EasyPlus Titrator Burette, 50 mL

Mettler Toledo | Mfr: 30043902| Item: EW-94190-58

Enhance the versatility of your Mettler Toledo titrators

$388.00USD / Each

$388.00USD / Each

Buy It Together

Current Item

Mettler Toledo EasyPlus Titrator Burette, 50 mL
Mettler Toledo EasyPlus Titrator Burette, 50 mL
9419058

$388.00USD / Each
+ + =
Total Price 19413.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Burette fits all EasyPlus titrators for extra volume applications
  • 1 year warranty

More About this Item

Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.

1 Year Warranty
Specifications
  • Capacity (mL)50
  • DescriptionEasyPlus Titrator Burette, 50 mL
  • Warranty1 year
Reviews
Q&A