Mettler Toledo StarterPac On-Site Installation with ACC for LA Analytical and Precision Toploading Balances
Representative image only.
Mettler Toledo StarterPac On-Site Installation with ACC for LA Analytical and Precision Toploading Balances
Mettler Toledo | Mfr: B99474130131| Item: EW-02629-68
Maximize your productivity and ensure compliance
$474.15USD / Each
$474.15USD / Each
Product Description
Key Features
  • NIST traceable
  • Professionally executed installation
More About this Item
StarterPac Installation provides documented installation by a factory-trained technician, and a certificate with NIST traceability. A professionally executed installation and qualification is essential in achieving the best results from your balance. These services ensure you peace of mind that your balance performs perfectly from the start.
Specifications
  • Compatible WithLA Analytical and Precision Toploading Balances
  • DescriptionStarterPac On-Site Installation with ACC for LA Analytical and Precision Toploading Balances