Novasina 111 1001 eVC-21 Filter for Acid and Chemical, for LabTouch and LabMaster Water Instruments
Representative image only.
Novasina 111 1001 eVC-21 Filter for Acid and Chemical, for LabTouch and LabMaster Water Instruments
Novasina | Mfr: 111 1001| Item: EW-37910-32
A number of filters are available to protect the measurement cell from abrasion or corrosive vapors
$611.00USD / Each
$611.00USD / Each
Buy It Together
Current Item
Novasina 111 1001 eVC-21 Filter for Acid and Chemical, for LabTouch and LabMaster Water Instruments
Novasina 111 1001 eVC-21 Filter for Acid and Chemical, for LabTouch and LabMaster Water Instruments
3791032
$611.00USD / Each
+ =
Total Price 1083.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sensor filters protect against contamination and errors
  • Chemical filter to eliminate the effect of such as acetic acid, sulfuric acid, lactic acid chlorine and more
  • 1 year warranty
More About this Item
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
1 Year Warranty
Specifications
  • DescriptioneVC-21 Filter for Acid and Chemical, for LabTouch and LabMaster Water Instruments