Novasina 111 1003 eVC-26 Filter for Oil and Dust, for LabTouch and LabMaster Water Activity Instruments
Representative image only.
Novasina 111 1003 eVC-26 Filter for Oil and Dust, for LabTouch and LabMaster Water Activity Instruments
Novasina | Mfr: 111 1003| Item: EW-37910-33
A number of filters are available to protect the measurement cell from abrasion or corrosive vapors
$325.00USD / Each
$325.00USD / Each
Buy It Together
Current Item
Novasina 111 1003 eVC-26 Filter for Oil and Dust, for LabTouch and LabMaster Water Activity Instruments
Novasina 111 1003 eVC-26 Filter for Oil and Dust, for LabTouch and LabMaster Water Activity Instruments
3791033
$325.00USD / Each
+ =
Total Price 797.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sensor filters protect against contamination and errors
  • Universal Filter agiast oil vapours, dust, amines, aromas, chloroform, and more
  • 1 year warranty
More About this Item
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
1 Year Warranty
Specifications
  • DescriptioneVC-26 Filter for Oil and Dust, for LabTouch and LabMaster Water Activity Instruments