Novasina 111 7212 Water Activity Meter ReDOx Filter for Alcohols and Aromas
Representative image only.
Novasina 111 7212 Water Activity Meter ReDOx Filter for Alcohols and Aromas
Novasina | Mfr: 111 7212| Item: EW-37910-34
A number of filters are available to protect the measurement cell from abrasion or corrosive vapors
$503.00USD / Each
$503.00USD / Each
Buy It Together
Current Item
Novasina 111 7212 Water Activity Meter ReDOx Filter for Alcohols and Aromas
Novasina 111 7212 Water Activity Meter ReDOx Filter for Alcohols and Aromas
3791034
$503.00USD / Each
+ =
Total Price 975.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sensor filters protect against contamination and errors
  • Eliminates effect of ethanol, glycols, propanols, methanols, yeast aromas, ketones, coffee aromas and more
  • 1 year warranty
More About this Item
For LabTouch and LabMaster Water Activity Instruments.
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
1 Year Warranty
Specifications
  • DescriptionWater Activity Meter ReDOx Filter for Alcohols and Aromas