Novasina ClimMate Replacement Humidity Adapter SAL-SC for nSens Probe
Representative image only.
Novasina ClimMate Replacement Humidity Adapter SAL-SC for nSens Probe
Novasina | Mfr: 260 1143| Item: EW-37910-63
Keep your Novasina ClimMate Portable Water Activity Meter producing optimal results
$108.25USD / Each
$108.25USD / Each
Buy It Together
Current Item
Novasina ClimMate Replacement Humidity Adapter SAL-SC for nSens Probe
Novasina ClimMate Replacement Humidity Adapter SAL-SC for nSens Probe
3791063
$108.25USD / Each
+ =
Total Price 580.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a wide range of accessories including humidity standards, probes, and more
More About this Item
These replacement accessories are compatible with Novasina ClimMate Portable Water Activity meter, model 37910-38. Meter sold separately.
This plastic adapter is suitable for use with nSens-xx probes with a 13 mm diameter. Use this adapter for humidity standards as a reduction piece and radial seal on the circumference of the respective sensor.
This adapter is compatible with the probes suitable with ClimMate water activity meter, model 37910-38, meter and probes sold separately.
Cole-Parmer cannot sell this product in China, Japan, Germany, France, Italy, Netherlands, Belgium, Great Britain, Spain, India, South Korea, Vietnam, Taiwan, Colombia or Brazil
Specifications
  • Compatible WithClimMate Portable Water Activity (aW) Meter 37910-38
  • DescriptionClimMate Replacement Humidity Adapter SAL-SC for nSens Probe