NSI Lab Solutions CMPT-063 Quantitative Pesticides in Edible Proficiency Test with Spiking Solution; 2/PK
Representative image only.
NSI Lab Solutions CMPT-063 Quantitative Pesticides in Edible Proficiency Test with Spiking Solution; 2/PK
NSI Lab Solutions | Item: UX-61204-68
Ensure state and accreditation regulation compliance with proficiency testing
$1,026.00USD / Pkg of 2
$1,026.00USD / Pkg of 2
Product Description
Includes
Two vials of blank matrix and one spiking solution.
Key Features
  • Designed for quantitative determination
  • Two studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • Sample set supplied as blank edible matrix and certified spiking solution
  • Adequate matrix and spiking solution for two analyses
  • At least 25% of BCC pesticide list will be present per study
More About this Item
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.
Specifications
  • Packaging2/PK
  • Product TypeHemp Chemistry Proficiency Test
  • Shelf Life Months (months)36
  • Shipping requirementFrozen ice packs
  • Storage requirementRefrigerate between 2 and 8°C
  • DescriptionQuantitative Pesticides in Edible Proficiency Test with Spiking Solution; 2/PK