NSI Lab Solutions FM-733-20 Multi-Organism CRM with Yeast/Mold; 20/PK
Representative image only.
NSI Lab Solutions FM-733-20 Multi-Organism CRM with Yeast/Mold; 20/PK
NSI Lab Solutions | Item: UX-05002-75
Multi-organism Certified Reference Material (CRM) for food safety testing
$345.00USD / Pkg of 20
$345.00USD / Pkg of 20
Product Description
Key Features
  • Compatible with 3M™ Petrifilm™ plate
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than 2 passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
  • Seven indicator test parameters with a single lyophilized sample: APC, total coliform, E. coli, Enterobacteriaceae, S. aureus, yeast, and mold
  • Pack of 20
More About this Item
Use these microbiological reference materials for quality control of food and agriculture samples. Each dehydrated pellet packaged in a vacuum-sealed glass vial contains multiple microorganisms that can be tested at once. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. These CRMs are designed for use with Petrifilm™ plate, FDA Bacteriological Analytical Manual (BAM) methods, or other re-hydratable film methods. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
3M™ and Petrifilm™ are trademarks of 3M.
Specifications
  • OrganismMultiple
  • Packaging20/PK
  • Product TypeFood and Agriculture Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • DescriptionMulti-Organism CRM with Yeast/Mold; 20/PK