NSI Lab Solutions QCI-146B Perchlorate Wastewater PT Express™; 2/PK
Representative image only.
NSI Lab Solutions QCI-146B Perchlorate Wastewater PT Express™; 2/PK
NSI Lab Solutions | Item: UX-61203-40
On-demand proficiency test (PT) for when you need results fast
$109.00USD / Pkg of 2
$109.00USD / Pkg of 2
Product Description
Key Features
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • Supplied in duplicate for convenience and value
  • Two 5.0-mL concentrate set for determination of Perchlorate
  • Each ampule produces 5 L of sample
  • Formulated in the range of 4.0 to 20 µg/L
More About this Item
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.
Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Specifications
  • Element/IonPerchlorate
  • FormLiquid
  • Volume (mL)5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionPerchlorate Wastewater PT Express™; 2/PK