Pomona Electronics 3789 Male Minigrabber Test Clips/Bnc Length 7in /177.8 Mm)

Representative image only.

Pomona Electronics 3789 Male Minigrabber Test Clips/Bnc Length 7in /177.8 Mm)

Pomona Electronics | Mfr: 3789| Item: EW-20009-26

Available for sale only in the United States and Canada

$26.10USD / Each
$29.00USD / Each

$26.10USD / Each
$29.00USD / Each
Product Description

Key Features

  • 3 month warranty

More About this Item

20 AWG Leads / BNC

3 Month Warranty
Specifications
  • DescriptionMale Minigrabber Test Clips/Bnc Length 7in /177.8 Mm)
  • Warranty3 month
Reviews
Q&A