Pomona Electronics 4970 Male 20 Awg Leads/Bnc (Wire Leng 177.8MM)

Representative image only.

Pomona Electronics 4970 Male 20 Awg Leads/Bnc (Wire Leng 177.8MM)

Pomona Electronics | Mfr: 4970| Item: EW-20009-33

Available for sale only in the United States and Canada

$17.54USD / Each
$19.49USD / Each

$17.54USD / Each
$19.49USD / Each
Product Description

Key Features

  • 3 month warranty

More About this Item

20 AWG Leads / BNC

3 Month Warranty
Specifications
  • DescriptionMale 20 Awg Leads/Bnc (Wire Leng 177.8MM)
  • Warranty3 month
Reviews
Q&A