Pomona Electronics 72902-2 Micro SMD Grabber Test Clips, Red
Representative image only.
Pomona Electronics 72902-2 Micro SMD Grabber Test Clips, Red
Pomona Electronics | Mfr: 72902-2| Item: EW-20009-63
Available for sale only in the United States and Canada
$24.30USD / Each
$27.00USD / Each
$24.30USD / Each
$27.00USD / Each
Product Description
Key Features
  • Designed for easy attachment to SMD chips with pitches down to 0.008"
  • 3 month warranty
More About this Item
Test Clips
Micro SMD Grabber Test Clips (PM729020 and PM729022)
Thin yet durable design permits attachment with close spacing in tight spaces. Insulated tip minimizes shorting on adjacent pins. Gold plating for low resistance connections. Holding rods available to reinforce connection positions during side-by-side measurements.
Available for sale in the United States and Canada only.
3 Month Warranty
Specifications
  • DescriptionMicro SMD Grabber Test Clips, Red