Pomona Electronics 72906-0 Mini Smd Grabber, Black, Test Clip
Representative image only.
Pomona Electronics 72906-0 Mini Smd Grabber, Black, Test Clip
Pomona Electronics | Mfr: 72906-0| Item: EW-20009-66
Available for sale only in the United States and Canada
$17.54USD / Each
$19.49USD / Each
$17.54USD / Each
$19.49USD / Each
Product Description
Key Features
  • Designed for easy attachment to SMD chips with pitches down to 0.032"
  • 3 month warranty
More About this Item
Test Clips
Mini SMD Grabber Test Clips
Insulated shaft can be bent up to 36 degrees for versatile attachment in awkward locations or close proximity to other clips. Rotating jaws increase versatility in connection and stacking. Thin (3mm) profile allows close placement to adjacent clips.
Available for sale in the United States and Canada only.
3 Month Warranty
Specifications
  • DescriptionMini Smd Grabber, Black, Test Clip