Pomona Electronics Solar MC4 to 4 mm Test Lead Set
Representative image only.
Pomona Electronics Solar MC4 to 4 mm Test Lead Set
Pomona Electronics | Mfr: PVLEAD1| Item: EW-20051-06
Connect to test tools or clamp meters with these photovoltaic test leads
$59.40USD / Each
$66.00USD / Each
$59.40USD / Each
$66.00USD / Each
Product Description
Includes
Set of black and red MC4 to 4 mm adapter test leads for solar applications.
Key Features
  • Ensures safe measurements on photovoltaic (PV) modules and systems
  • Connect to instruments that accept 4 mm sheathed banana plugs
  • Nickel plated contacts
  • 1 year warranty
More About this Item
Monitor solar photovoltaic (PV) systems in solar power plants with these test lead sets. Users can connect to test tools or clamp meters with two different options.
Connect measuring devices to PV power plants, for use in regular tests, measurements, and troubleshooting on solar PV panels. The black lead is an MC4 (female) to 4 mm sheathed banana plug on a 60-inch test lead. The red lead is an MC4 (male) to 4 mm sheathed banana plug on a 60-inch test lead.
1 Year Warranty
Specifications
  • DescriptionSolar MC4 to 4 mm Test Lead Set