Qorpak 2911 Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs

Representative image only.

Qorpak 2911 Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs

Qorpak | Mfr: PAP-03114| Item: EW-06148-21

Meet government regulations for mailing liquids

$219.50USD / Case of 24

$219.50USD / Case of 24
Product Description

More About this Item

Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps.

Specifications
  • Fits Bottle / Beaker Size (mL)60
  • MaterialFiberboard
  • Cap MaterialMetal
  • AutoclavableNo
  • Outside Diameter (OD) (in)1 3/4
  • Outside Diameter (OD) (cm)4.4
  • Overall Height (in)4 1/2
  • Overall Height (cm)11.4
  • DescriptionBottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs
Reviews
Q&A