Qorpak 2912 Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs

Representative image only.

Qorpak 2912 Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs

Qorpak | Mfr: PAP-03115| Item: EW-06148-31

Meet government regulations for mailing liquids

$235.25USD / Case of 24

$235.25USD / Case of 24
Product Description

More About this Item

Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps.

Specifications
  • Fits Bottle / Beaker Size (mL)125
  • MaterialFiberboard
  • Cap MaterialMetal
  • AutoclavableNo
  • Outside Diameter (OD) (in)2
  • Outside Diameter (OD) (cm)5.1
  • Overall Height (in)5 1/2
  • Overall Height (cm)14
  • DescriptionBottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs
Reviews
Q&A