Reagents Acetic Acid, 10% (v/v), Standardized, Natural Poly Bottle, 1 L

Representative image only.

Reagents Acetic Acid, 10% (v/v), Standardized, Natural Poly Bottle, 1 L

Reagents | Mfr: CA008900-1A| Item: EW-78914-76

Manufactured using the highest quality raw materials in ISO 9001 facility

$65.00USD / Each

$65.00USD / Each
Product Description

Key Features

  • 1:9
  • Assay 1.715 to 1.785 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1.01
  • Boiling point: approximately 118 °C (244.4 °F)
  • Melting poing: approximately 0 °C (32 °F)
  • Flash point: 39 °C (102.2 °F)
  • Solubility information: miscible
  • pH range: 2.3
  • 1 L natural poly bottle
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

3 Year Warranty
Specifications
  • Concentration10% v/v
  • ClassStandardized
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly bottle
  • DOT InformationACETIC ACID SOLUTION, 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN2790
  • Shelf Life (days)1095
  • Shelf Life (months)36
  • DescriptionAcetic Acid, 10% (v/v), Standardized, Natural Poly Bottle, 1 L
  • Warranty3 year
Documents & Certificates
Reviews
Q&A