Reagents Acetic Acid, Glacial, 2.5L, Purified

Representative image only.

Reagents Acetic Acid, Glacial, 2.5L, Purified

Reagents | Mfr: C5100500-2.5D1| Item: UX-78920-15

Manufactured using the highest quality raw materials in ISO 9001 facility

$124.00USD / Each

$124.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 5 year warranty

More About this Item

Glacial Acetic Acid, 500mL,Purified Grade, 64-19-7 CAS Number, CH3COOH Chemical Formula, 60.05g/mol Molecular Weight, Colorless, Liquid, 118°C Boiling Point, 16.64°C Melting Point, 39°C Flash Point, <2 pH Range, 1.05 Density, 1.37nD Refractive Index, 60 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaCH3COOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight60.05
  • Vapor Pressure (torr)11.4
  • PackagingClear Glass Bottle
  • DOT InformationACETIC ACID, GLACIAL, 8 (3)
  • CAS Number64-19-7
  • UN NumberUN2789
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionAcetic Acid, Glacial, 2.5L, Purified
  • Warranty5 years
Documents & Certificates
Reviews
Q&A