Reagents® Acetic Acid, Glacial, Purified
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Reagents and solutions to meet your lab and research needs
Glacial Acetic Acid, 500mL,Purified Grade, 64-19-7 CAS Number, CH3COOH Chemical Formula, 60.05g/mol Molecular Weight, Colorless, Liquid, 118°C Boiling Point, 16.64°C Melting Point, 39°C Flash Point, <2 pH Range, 1.05 Density, 1.37nD Refractive Index, 60 Months Shelf Life.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$124.00 - $546.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Acetic Acid, Glacial, 2.5L, Purified EW-78920-15 Mfr # C5100500-2.5D1
COMPARE
Acid 2500 Acetic Acid, Glacial, 2.5L, Purified
$124.00USD / Each
Reagents Acetic Acid, Glacial, 20L, Purified EW-78928-77 Mfr # C5100500-20E7
COMPARE
Acid 20000 Acetic Acid, Glacial, 20L, Purified
$546.00USD / Each