Reagents Acetic Acid, Glacial, 500mL, ACS Reagent; Amber Glass Container

Representative image only.

Reagents Acetic Acid, Glacial, 500mL, ACS Reagent; Amber Glass Container

Reagents | Mfr: C5100600-500C| Item: EW-78910-09

Manufactured using the highest quality raw materials in ISO 9001 facility

$94.10USD / Each

$94.10USD / Each

Buy It Together

Current Item

Reagents Acetic Acid, Glacial, 500mL, ACS Reagent; Amber Glass Container
Reagents Acetic Acid, Glacial, 500mL, ACS Reagent; Amber Glass Container
7891009

$94.10USD / Each
+ =
Total Price 151.40 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Minimum Assay 99.7%
  • 3 year warranty

More About this Item

Glacial Acetic Acid, 500mL, ACS Reagent Grade, 64-19-7 CAS Number, CH3COOH Chemical Formula, 60.05g/mol Molecular Weight, Colorless, Liquid, 118°C Boiling Point, 16.64°C Melting Point, 39°C Flash Point, <2 pH Range, 1.05 Density, 1.37nD Refractive Index, 36 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

3 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaCH3COOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight60.05
  • Vapor Pressure (torr)11.4
  • PackagingGlass Amber Container
  • DOT InformationACETIC ACID, GLACIAL, 8 (3)
  • UN NumberUN2789
  • CAS Number64-19-7
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionAcetic Acid, Glacial, 500mL, ACS Reagent
  • Warranty3 year
Documents & Certificates
Reviews
Q&A