Reagents Acetone, Meets ASTM D 329, Metal Can, 20 L

Representative image only.

Reagents Acetone, Meets ASTM D 329, Metal Can, 20 L

Reagents | Mfr: C1102550-20E6| Item: EW-78927-79

Manufactured using the highest quality raw materials in ISO 9001 facility

$664.00USD / Each

$664.00USD / Each
Product Description

Key Features

  • ASTMD 329 certified
  • Colorless liquid
  • Flash point: -20 °C (-4 °F)
  • Density: 0.8
  • 20 L metal can
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeASTM
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • Synonyms2-Propanone, Dimethyl ketone
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight58.08
  • Vapor Pressure (torr)176.7644
  • PackagingMetal Can
  • DOT InformationACETONE, 3
  • Storage SymbolAmbient
  • CAS Number67-64-1
  • UN NumberUN1090
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, Meets ASTM D 329, Metal Can, 20 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A